Copper(II) trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1292-25G T1292-5G
TCT1292-25GEA 98.1 USD
TCT1292-25G TCT1292-5G
Copper(II) trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Copper(II) trifluoromethanesulphonate

CAS Number: 34946-82-2
MDL Number: MFCD00077492
Molecular Formula: CHF3O3S
Molecular Weight: 361.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Glycosylation, Stable Lewis Acids in Aqueous Media
Other Product Names / Synonyms: Copper(II) Triflate, Trifluoromethanesulfonic Acid Copper(II) Salt

Formula: C₂CuF₆O₆S₂
MDL Number: MFCD00077492
CAS Number: 34946-82-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6af86c5054c9f7fdfc2279422743e7a6f70a26b&vid=d6af86c5054c9f7fdfc2279422743e7a6f70a26b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR