6-Chloronicotinoyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C1726-25G
C1726-5G
TCC1726-25GEA
108.51
USD
TCC1726-25G
TCC1726-5G
6-Chloronicotinoyl chloride ≥98.0% (by titrimetric analysis)
6-Chloronicotinoyl chloride
CAS Number: 58757-38-3
MDL Number: MFCD00051775
Molecular Formula: C6H3Cl2NO
Molecular Weight: 176.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 85
Melting point (°C): 51
Other Product Names / Synonyms: 6-Chloropyridine-3-carbonyl Chloride
Formula:
C₆H₃Cl₂NO Boiling Pt: 85 °C Melting Pt: 51 °C |
MDL Number:
MFCD00051775 CAS Number: 58757-38-3 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...