Anti-NK-1.1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: PK136]

Supplier: Biolegend
108731 108732 108741
108731-BLEA 217.3 USD
108731-BL 10756-888 10756-900
Anti-NK-1.1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: PK136]
Antibodies
Brilliant Violet 421™ anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC, IF; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: NK-1.1
Clonality: Monoclonal
Clone: PK136
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR