Anti-IgG Donkey Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20076 20076-1
71003-802EA 260.49 USD
71003-802 71003-804
Anti-IgG Donkey Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR