Paraformaldehyde 16% (w/v) in water methanol-free

Supplier: BeanTown Chemical


ampouled under argon

140770-10X10ML 140770-2X10ML
BT140770-10X10EA 98.07 USD
BT140770-10X10 BT140770-2X10M
Paraformaldehyde 16% (w/v) in water methanol-free
Paraformaldehyde

CAS: 30525-89-4; MDL No: MFCD00133991 Liquid; Linear Formula: (CH2O)n; MW: 30.03 (monomer)

Formula: (CH₂O)n
MDL Number: MFCD00133991
CAS Number: 30525-89-4
UN: 2213
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR