D-(-)-Tartaric acid ≥99%

Supplier: BeanTown Chemical

144655-100G 144655-25G 144655-500G
BT144655-100GEA 112.23 USD
BT144655-100G BT144655-25G BT144655-500G
D-(-)-Tartaric acid ≥99%
D-(-)-Tartaric acid

CAS: 147-71-7; EC No: 205-695-6; MDL No: MFCD00004238 Crystalline; Molecular Formula: C4H6O6; MW: 150.09 Melting Point: 172-174° Optical Rotation: [α]20/D -12°, c = 20 in water


  • Optical rotation: [α]20/D -12°, c = 20 in water
  • Crystalline

Formula: C₄H₆O₆
Melting Pt: 172…174 °C
MDL Number: MFCD00004238
CAS Number: 147-71-7
EINECS: 205-695-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR