Potassium carbonate, anhydrous ≥99%

Supplier: BeanTown Chemical

124060-10KG 124060-2.5KG 124060-500G
BT124060-10KGEA 247.39 USD
BT124060-10KG BT124060-2.5KG BT124060-500G
Potassium carbonate, anhydrous ≥99%
Potassium carbonate

CAS: 584-08-7; EC No: 209-529-3; MDL No: MFCD00011382; RTECS: TS7750000 Powder/Granular; Linear Formula: K2CO3; MW: 138.21 Melting Point: 891° Density (g/mL): 2.43 Hygroscopic


  • RTECS: TS7750000
  • Hygroscopic
  • Powder/Granular

Formula: K₂CO₃
Melting Pt: 891 °C
Density: 2.43 g/mL
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR