L(-)-Malic acid ≥99%, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.
Soluble in 100 mg/mL - Clear to Very Slightly Hazy, Colorless.
L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.
Formula:
C₄H₆O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00064213 CAS Number: 97-67-6 |
Specification Test Results
Synonyms | L-Hydroxysuccinic acid; L-Hydroxybutanedioic acid; (S)-( -)-2-Hydroxysuccinic acid |
Form | White Powder |
Storage | Store at Room Temperature (15-30 °C). |
Optical Rotation | (c=2, H2O) -1.5 to -3.0 ° |
pka | (1) 3.46, (2) 5.10 (25 °C)(Lit.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...