Human Amylin (20-29)
Supplier: BACHEM AMERICAS INC
4030350.0001 4030350.0005
H-3746.0001BAEA 147.06 USD
H-3746.0001BA H-3746.0005BA
Human Amylin (20-29)
Proteins and Peptides
Amylin (20-29) (or hIAPP 20-29) forms fibrils that are ultrastructurally identical to amyloid fibrils seen in pancreatic islets. The region SNNFGAILSS appears to be the most important amyloidogenic sequence of hIAPP (human islet amyloid polypeptide).