Chitin

Supplier: Thermo Scientific Chemicals

J61206-22 J61206-36
AAJ61206-22EA 77.12 USD
AAJ61206-22 AAJ61206-36
Chitin
Chitin

A long-chain polymer of a N-acetylglucosamine


Powder


Applications: A long-chain polymer of a N-acetylglucosamine

Formula: C₃₀H₅₀N₄O₁₉
Storage Temperature: Ambient
MDL Number: MFCD00466914
CAS Number: 1398-61-4
EINECS: 215-744-3
Merck Index: 14,2065

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR