Methyl (phenylthio)acetate
Supplier: TCI America
CAS Number: 17277-58-6
MDL Number: MFCD00039805
Molecular Formula: C9H10O2S
Molecular Weight: 182.24
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 102
Flash Point (°C): 110
Specific Gravity (20/20): 1.17
Other Product Names / Synonyms: (Phenylthio)acetic Acid Methyl Ester
Formula:
C₉H₁₀O₂S Boiling Pt: 102 °C Density: 1.17 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00039805 CAS Number: 17277-58-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...