3-(Methylthio)propionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Methylthio)propionic Acid

M0811-25G M0811-100G
TCM0811-25GEA 61.05 USD
TCM0811-25G TCM0811-100G
3-(Methylthio)propionic acid ≥98.0% (by GC, titration analysis)
3-(Methylthio)propionic acid

CAS Number: 646-01-5
MDL Number: MFCD00059635
Molecular Formula: C4H8O2S
Molecular Weight: 120.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 104
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: 3-(Methylmercapto)propionic Acid

Formula: C₄H₈O₂S
MW: 120.17 g/mol
Boiling Pt: 130 °C
Density: 1.16 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00059635
CAS Number: 646-01-5
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=429a0b2ae04e40db69c5211be17b7d47612e1633&vid=429a0b2ae04e40db69c5211be17b7d47612e1633&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR