5-Cyano-1H-imidazole-4-carboxamide hydrate ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
C1338-5G
C1338-25G
TCC1338-5GEA
32.54
USD
TCC1338-5G
TCC1338-025G
5-Cyano-1H-imidazole-4-carboxamide hydrate ≥95.0% (by HPLC, titration analysis)
5-Cyano-1H-imidazole-4-carboxamide hydrate
CAS Number: 5372-23-6
MDL Number: MFCD04038014
Molecular Formula: C5H4N4O
Molecular Weight: 136.11
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 278
Formula:
C₅H₄N₄O MW: 136.11 g/mol Melting Pt: 278 °C |
MDL Number:
MFCD04038014 CAS Number: 5372-23-6 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...